Specifications
- Antibody type:Primary
- Antigen name:RNA Binding Motif Protein 24
- Antigen symbol:RBM24
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:150 µL
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Fusion Protein
- Purification:Antigen affinity Purification
- Cat. no.:10093-406
- Supplier No.:18178-1-AP
Specifications
About this item
RNA-binding proteins have a pivotal role in post-transcriptionally regulating gene expression. During early development they exhibit temporally and spatially regulated expression pattern, and affect the proteome of a single cell by controlling alternative splicing, transport, localization, and stability of their target RNAs . Their expression are tissue-specific, and their crucial influence on the protein set-up especially in muscle cells has become recognized recently . RBM24 is one RNA-binding protein and plays a fundamental role in regulating cardiac gene expression, sarcomeric assembly, and cardiac contractility and may thus represent a novel pathway to cardiomyopathy .
IHC: Human Skeletal Muscle Tissue, 1:20-1:200
Type: Primary
Antigen: RBM24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat