Order Entry
Canada
ContactUsLinkComponent
Anti-CD3E Rabbit Polyclonal Antibody
Anti-CD3E Rabbit Polyclonal Antibody
Catalog # 10084-316
Supplier:  Proteintech
Anti-CD3E Rabbit Polyclonal Antibody
Catalog # 10084-316
Supplier:  Proteintech
Supplier Number:  17617-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD3 Molecule Epsilon
  • Antigen symbol:
    CD3E
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10084-316
  • Supplier No.:
    17617-1-AP

Specifications

About this item

CD3 is a complex of proteins that directly associates with the T cell receptor (TCR). The TCR/CD3 complex of T-lymphocytes consists of either a TCR alpha/beta or TCR gamma/delta heterodimer coexpressed at the cell surface with the invariant subunits of CD3 labeled gamma, delta, epsilon, zeta, and eta. The TCR recognizes antigens bound to major histocompatibility complex (MHC) molecules. TCR-mediated peptide-MHC recognition is transmitted to the CD3 complex, leading to the intracellular signal transduction. CD3 is considered to be a pan-T cell marker for detection of normal and neoplastic T cells.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Lymphoma Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: