Order Entry
Canada
ContactUsLinkComponent
Anti-NRSN2 Rabbit Polyclonal Antibody
Anti-NRSN2 Rabbit Polyclonal Antibody
Catalog # 10091-308
Supplier:  Proteintech
Anti-NRSN2 Rabbit Polyclonal Antibody
Catalog # 10091-308
Supplier:  Proteintech
Supplier Number:  17574-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurensin 2
  • Antigen symbol:
    NRSN2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10091-308
  • Supplier No.:
    17574-1-AP

Specifications

About this item

Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Brain, 1:20-1:200

Type: Primary
Antigen: NRSN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human