Order Entry
Canada
ContactUsLinkComponent
 
Anti-Inhibin beta A-Specific Rabbit Polyclonal Antibody
Catalog # 10088-900
Supplier:  Proteintech
undefined
Anti-Inhibin beta A-Specific Rabbit Polyclonal Antibody
Catalog # 10088-900
Supplier:  Proteintech
Supplier Number:  17524-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Inhibin Beta A
  • Antigen symbol:
    INHBA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Peptide
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10088-900
  • Supplier No.:
    17524-1-AP

Specifications

About this item

Inhibin βA(INHBA) is a ligand in the transforming growth factor β (TGF-β) superfamily. Inhibin βA forms a disulfide-linked homo-dimer known as activin A, which has been implicated in multiple biologic processes, including neoplastic progression. The inhibin βA can also bind the βB and α isoforms to form activin AB and inhibin, respectively.

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: Inhibin beta A-Specific
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat