Anti-PRPF40A Rabbit Polyclonal Antibody
Catalog # 10092-832
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Pre-mRNA Processing Factor 40A
- Antigen symbol:PRPF40A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10092-832
- Supplier No.:17392-1-AP
Specifications
About this item
Western Blot: HEK-293 Cells, 1:500-1:5000
Type: Primary
Antigen: PRPF40A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse