Order Entry
Canada
ContactUsLinkComponent
Anti-MRPS27 Rabbit Polyclonal Antibody
Anti-MRPS27 Rabbit Polyclonal Antibody
Catalog # 10090-480
Supplier:  Proteintech
Anti-MRPS27 Rabbit Polyclonal Antibody
Catalog # 10090-480
Supplier:  Proteintech
Supplier Number:  17280-1-AP-150UL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitochondrial Ribosomal Protein S27
  • Antigen symbol:
    MRPS27
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10090-480
  • Supplier No.:
    17280-1-AP

Specifications

About this item

Genes encoding PPR (pentatricopeptide repeat)-containing proteins constitute one of the largest gene families in plants. The majority of these proteins are predicted to target organelles and to bind to RNA. Strikingly, there is a dearth of these proteins in mammals, although genomic searches reveal six candidates, all of which are also predicted to target the mitochondrion. One of these proteins, MRPS27is participated in transcription and translation respectively. MRPS27is a component of the small subunit of the mitochondrial ribosome that is encoded by the nuclear genome .

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human Kidney, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: MRPS27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat