Anti-RECQL4 Rabbit Polyclonal Antibody
10093-492
- Antibody type:Primary
- Antigen name:RecQ Protein-like 4
- Antigen symbol:RECQL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10093-492
- Supplier No.:17008-1-AP
RECQ protein-like 4(RECOL4), which has helicase domains contain 7 consensus motifs in the middle, is a gene responsible for Werner syndorme and Bloom syndrome. It forms a stable complex with UBR1 adn UBR2, which has a DNA-stimulated ATPase activity. RECOL4 may involve in repairment of DNA damage by interaction with APE1 and FEN1, key participants in base excision repair. This is a rabbit polyclonal antibody raised against part of C-terminal of RECQ4 of human origin.
Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2 Cells, 1:20-1:200; IHC: Human Kidney Tissue, 1:20-1:200; IP: HeLa Cells, 1:500-1:5000
Type: Primary
Antigen: RECQL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human