Anti-TSR1 Rabbit Polyclonal Antibody
10096-450
- Antibody type:Primary
- Antigen name:TSR1 20S rRNA accumulation homolog
- Antigen symbol:TSR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10096-450
- Supplier No.:16887-1-AP
Western Blot: Mouse Liver Tissue, 1:500-1:5000
Type: Primary
Antigen: TSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat