Anti-AMN1 Rabbit Polyclonal Antibody
Catalog # 10082-842
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Antagonist Of Mitotic Exit Network 1 Homolog
- Antigen symbol:AMN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10082-842
- Supplier No.:14694-1-AP
Specifications
About this item
IHC: Human Colon cancer Tissue, 1:20-1:200; Western Blot: Human Colon Tissue, 1:200-1:1000
Type: Primary
Antigen: AMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat