Anti-TERF2IP Rabbit Polyclonal Antibody
10095-790
- Antibody type:Primary
- Antigen name:Telomeric Repeat Binding Factor 2, Interacting Protein
- Antigen symbol:TERF2IP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10095-790
- Supplier No.:14595-1-AP
Western Blot: HEK-293 Cells, 1:1000-1:10000; IF: Hela Cells, 1:10-1:100; IHC: Human tonsillitis Tissue, 1:20-1:200; IP: HeLa Cells, 1:200-1:2000
Type: Primary
Antigen: TERF2IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat