Anti-TCF7L1 Rabbit Polyclonal Antibody
Catalog # 10095-728
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Transcription Factor 7-like 1
- Antigen symbol:TCF7L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:TCF-3|TCF3
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10095-728
- Supplier No.:14519-1-AP
Specifications
About this item
TCF7L1 is one of TCF/LEF transcription factors that are mediators of the Wnt signaling pathway and are antagonized by the TGF-beta signaling pathway. It binds to DNA and acts as a repressor in the absence of CTNNB1, and as an activator in its presence. It's necessary for the terminal differentiation of epidermal cells, the formation of keratohyalin granules and the development of the barrier function of the epidermis
Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human breast cancer, 1:20-1:200
Type: Primary
Antigen: TCF7L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat