Order Entry
Canada
ContactUsLinkComponent
Anti-MTMR2 Rabbit Polyclonal Antibody
Anti-MTMR2 Rabbit Polyclonal Antibody
Catalog # 10090-568
Supplier:  Proteintech
undefined
Anti-MTMR2 Rabbit Polyclonal Antibody
Catalog # 10090-568
Supplier:  Proteintech
Supplier Number:  14372-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Myotubularin Related Protein 2
  • Antigen symbol:
    MTMR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    MTMR2_HUMAN|Myotubularin related protein 2|KIAA1073|MTMR2|Myotubularin-related protein 2.|CMT4B1|CMT4B
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10090-568
  • Supplier No.:
    14372-1-AP

Specifications

About this item

MTMR2(myotubularin-related protein 2) is also named as KIAA1073 and belongs to the non-receptor class myotubularin subfamily, which is characterized by the presence of a phosphatase domain. It may regulate vesicular trafficking in Schwann cells and its reversible phosphorylation represents a critical mechanism for regulating the subcellular localization of MTMR2. Mtmr2 is a 73 kDa protein in its monomeric form and exist as a dimer within cells and the protein re-localizes to membranes in hypo-osmotic settings. Defects in MTMR2 are the cause of Charcot-Marie-Tooth disease type 4B1 (CMT4B1).

Western Blot: Human cerebellum Tissue, 1:500-1:5000

Type: Primary
Antigen: MTMR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat