Anti-ECEL1 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The M13 family has seven members, including ECEL1 (endothelin-converting enzyme-like 1), one of the newest members. ECEL1 is expressed predominantly in the central nervous system. It has been proposed that the enzyme has a role in the nervous regulation of the respiratory system.It may contribute to the degradation of peptide hormones and be involved in the inactivation of neuronal peptides.It has 2 isoforms produced by alternative splicing and 3 glycosylation sites.It can be detected a band of 95kDa in CHO cells by western blot because of the heavily glycosylated.
Type: Primary
Antigen: ECEL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Size:150 µL
- Clonality:Polyclonal
- Purification:Antigen affinity Purification
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Shipping temperature:Store at -20°C
- Antigen name:Endothelin Converting Enzyme-Like 1
- Cat. No.:10086-240
- Reactivity:Human|Rat|Mouse
- Immunogen:Recombinant Protein
- Conjugation:Unconjugated
- Antigen symbol:ECEL1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



