Anti-LEFTY2 Rabbit Polyclonal Antibody
10089-590
- Antibody type:Primary
- Antigen name:Left-right determination Factor 2
- Antigen symbol:LEFTY2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:TGFB4|LEFTA|LEFTYA|EBAF
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10089-590
- Supplier No.:13991-1-AP
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human endometrial cancer, 1:20-1:200
Type: Primary
Antigen: LEFTY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat