Anti-Trefoil factor 1 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
TFF1 (formerly known as pS2) is a small-secreted protein that belongs to the family of trefoil peptides, and characterized by the presence of one to six cysteine-rich P domains. TFF1 expression is frequently lost in gastric carcinoma, probably through mechanism of DNA methylation, and it is therefore considered as a tumor suppressor gene. The function of TFF1 was first controversially considered as an oncogene in breast cancer.
Western Blot: Human Stomach Tissue, 1:200-1:2000; IHC: Human Stomach cancer Tissue, 1:20-1:200
Type: Primary
Antigen: Trefoil factor 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Size:150 µL
- Western blot:Yes
- Clonality:Polyclonal
- Purification:Antigen affinity Purification
- Antibody type:Primary
- Isotype:IgG
- Host:Rabbit
- Shipping temperature:Store at -20°C
- Antigen name:Trefoil Factor 1
- Cat. No.:10092-864
- Reactivity:Human
- Immunogen:Recombinant Protein
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:TFF1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- ELISA:Yes
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



