Anti-LSS Rabbit Polyclonal Antibody
Catalog # 10089-638
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Lanosterol Synthase (2 3-oxidosqualene-lanosterol cyclase)
- Antigen symbol:LSS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:OSC|CTRCT44
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10089-638
- Supplier No.:13715-1-AP
Specifications
About this item
LSS, also named as OSC, is lanosterol synthase which catalyzes the cyclization of (S)-2,3 oxidosqualene to lanosterol, a reaction that forms the sterol nucleus. LSS gene has three isoforms with MW 74 kDa and 82-85 kDa. This antibody can recognize all these three isoforms.
Western Blot: Mouse Testis Tissue, 1:500-1:5000; IP: HepG2 Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200; IF: HepG2 Cells, 1:20-1:200
Type: Primary
Antigen: LSS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat