Specifications
- Antibody type:Primary
- Antigen name:Sperm Autoantigenic Protein 17
- Antigen symbol:SPA17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:CT22|SP17|SP17-1
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10095-036
- Supplier No.:13367-1-AP
Specifications
About this item
Sperm surface protein Sp17(SPA17)is a 151 amino acids protein (17kDa), which is a sperm surface peripheral membrane protein. It is a highly conserved mammalian protein expressed in developing spermatozoa and mature spermatids, and reported to play a role in the interaction of sperm with the zona pellucida. SP17 exists as a homodimer and localizes to the head and tail of spermatozoa. SP17 has been identified as a cancer/testis antigen and is expressed in ovarian cancer and multiple myeloma. This suggests that SP17 could be suitable as a target in tumor immunotherapy.
Western Blot: Mouse Testis Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IP: Mouse Testis Tissue, 1:200-1:1000
Type: Primary
Antigen: SPA17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat