Anti-TRIB3 Rabbit Polyclonal Antibody
Catalog # 10096-310
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Tribbles Pseudokinase 3
- Antigen symbol:TRIB3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:Neuronal cell death inducible putative kinase|SKIP3|TRIB3
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10096-310
- Supplier No.:13300-1-AP
Specifications
About this item
Tribbles homolog 3 (TRIB3) is a pseudokinase involved in the regulation of several signaling pathways involved in cell survival and/or cell stress.Whereas mRNA levels of TRIB3 are related with a poor prognosis, TRIB3 protein is associated with a good prognosis in human breast cancer patients, possibly due to the fact that TRIB3 is involved in hypoxia tolerance.
Western Blot: Mouse Liver Tissue, 1:500-1:5000
Type: Primary
Antigen: TRIB3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat