Order Entry
Canada
ContactUsLinkComponent
Anti-NGEF Rabbit Polyclonal Antibody
Anti-NGEF Rabbit Polyclonal Antibody
Catalog # 10091-086
Supplier:  Proteintech
undefined
Anti-NGEF Rabbit Polyclonal Antibody
Catalog # 10091-086
Supplier:  Proteintech
Supplier Number:  13271-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neuronal Guanine Nucleotide Exchange Factor
  • Antigen symbol:
    NGEF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    ARHGEF27|EPHEXIN
  • Storage buffer:
    PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10091-086
  • Supplier No.:
    13271-1-AP

Specifications

About this item

Western Blot: Mouse Brain Tissue, 1:200-1:2000

Type: Primary
Antigen: NGEF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat