Anti-FCAR Rabbit Polyclonal Antibody
10084-392
- Antibody type:Primary
- Antigen name:Fc fragment of IgA receptor
- Antigen symbol:FCAR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:FCAR_HUMAN|Immunoglobulin alpha Fc receptor.|Fc alpha receptor|CD89|IgA Fc receptor|FCAR
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10084-392
- Supplier No.:13245-1-AP
Western Blot: HL-60 Cells, 1:200-1:1000
Type: Primary
Antigen: CD89
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human