Order Entry
Canada
ContactUsLinkComponent
Anti-CD247 Rabbit Polyclonal Antibody
Anti-CD247 Rabbit Polyclonal Antibody
  10084-304
Anti-CD247 Rabbit Polyclonal Antibody
  10084-304
 :  12837-2-AP-150UL

 

  • Antibody type:
    Primary
  • Antigen name:
    CD247 molecule
  • Antigen symbol:
    CD247
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    Tcrz|Cd3-zeta|Cd3h|Cd3zeta|Tcrk|Cd3-eta|T3z|Cd3z|Cd3|AW552088
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10084-304
  • Supplier No.:
    12837-2-AP

 

 

CD247, also known as CD3Z (T-cell surface glycoprotein CD3 zeta chain), belongs to the CD3Z/FCER1G family. CD3 is a complex of proteins that directly associates with the T cell receptor (TCR). The TCR/CD3 complex of T-lymphocytes consists of either a TCR alpha/beta or TCR gamma/delta heterodimer coexpressed at the cell surface with the invariant subunits of CD3 labeled gamma, delta, epsilon, zeta, and eta. The TCR recognizes antigens bound to major histocompatibility complex (MHC) molecules. TCR-mediated peptide-MHC recognition is transmitted to the CD3 complex, leading to the intracellular signal transduction. CD3 is considered to be a pan-T cell marker for detection of normal and neoplastic T cells.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Lymphoma Tissue, 1:20-1:200; FC:Jurkat Cells, N/A

Type: Primary
Antigen: CD247
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat