Anti-STK38L Rabbit Polyclonal Antibody
Catalog # 10095-322
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Serine/Threonine Kinase 38-Like
- Antigen symbol:STK38L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:KIAA0965|NDR2|NDR2 protein kinase|Nuclear Dbf2-related kinase 2
- Storage buffer:PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10095-322
- Supplier No.:12697-1-AP
Specifications
About this item
Western Blot:A375 Cells, 1:200-1:2000
Type: Primary
Antigen: STK38L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat