Anti-GSTM1 Rabbit Polyclonal Antibody
10087-926
- Antibody type:Primary
- Antigen name:Glutathione S-transferase Mu 1
- Antigen symbol:GSTM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:GTM1|GSTM1b-1b|MU-1|GSTM1a-1a|GTH4|GST1|EC 2.5.1.18|H-B|MU|MGC26563|GSTM1-1
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10087-926
- Supplier No.:12412-1-AP
Western Blot: Mouse ovary Tissue, 1:200-1:2000; IHC: Human gliomas, 1:20-1:200
Type: Primary
Antigen: GSTM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse