Anti-LBR Rabbit Polyclonal Antibody
Catalog # 10089-544
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Lamin B Receptor
- Antigen symbol:LBR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:PHA|TDRD18|LMN2R|DHCR14B
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10089-544
- Supplier No.:12398-1-AP
Specifications
About this item
Western Blot:A375 Cells, 1:1000-1:10000; IF: HepG2, 1:10-1:100
Type: Primary
Antigen: LBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat