Anti-CLIC4 Rabbit Polyclonal Antibody
Catalog # 10084-864
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Chloride Intracellular Channel 4
- Antigen symbol:CLIC4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:H1|huH1|p64H1|CLIC4L|MTCLIC
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10084-864
- Supplier No.:12298-2-AP
Specifications
About this item
Western Blot: Mouse Kidney Tissue, 1:200-1:2000
Type: Primary
Antigen: CLIC4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: