Order Entry
Canada
ContactUsLinkComponent
Anti-SSSCA1 Rabbit Polyclonal Antibody
Anti-SSSCA1 Rabbit Polyclonal Antibody
Catalog # 10095-210
Supplier:  Proteintech
undefined
Anti-SSSCA1 Rabbit Polyclonal Antibody
Catalog # 10095-210
Supplier:  Proteintech
Supplier Number:  12273-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sjogren Syndrome/Scleroderma Autoantigen 1
  • Antigen symbol:
    SSSCA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    p27|ALURBP
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10095-210
  • Supplier No.:
    12273-1-AP

Specifications

About this item

Western Blot: HeLa Cells, 1:200-1:2000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: SSSCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat