Anti-STK3 Rabbit Polyclonal Antibody
Catalog # 10095-308
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Serine/Threonine Kinase 3
- Antigen symbol:STK3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:MESS1|Mammalian STE20-Like Protein Kinase 2 |KRS1|EC 2.7.11|MST-2|MST2
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10095-308
- Supplier No.:12097-1-AP
Specifications
About this item
Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human ovary tumor Tissue, 1:20-1:200; IF: Hela Cells, 1:20-1:200
Type: Primary
Antigen: STK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat