Anti-VDAC2 Rabbit Polyclonal Antibody
10081-330
- Antibody type:Primary
- Antigen name:Voltage-dependent Anion Channel 2
- Antigen symbol:VDAC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10081-330
- Supplier No.:11663-1-AP
Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human prostate cancer, 1:20-1:200
Type: Primary
Antigen: VDAC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat