Order Entry
Canada
ContactUsLinkComponent
Anti-ZNF266 Rabbit Polyclonal Antibody
Anti-ZNF266 Rabbit Polyclonal Antibody
Catalog # 10097-328
Supplier:  Proteintech
undefined
Anti-ZNF266 Rabbit Polyclonal Antibody
Catalog # 10097-328
Supplier:  Proteintech
Supplier Number:  11374-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Zinc Finger Protein 266
  • Antigen symbol:
    ZNF266
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10097-328
  • Supplier No.:
    11374-1-AP

Specifications

About this item

ZNF266, also named as HZF1, was expressed in ubiquitous tissues and various hematopoietic cell lines. Expression of ZNF266 in K562 cells was increased after being induced by hemin or phorbol myristate acetate (PMA), suggesting that HZF1 play important roles in erythroid and megakaryocytic differentiation.

Western Blot: HeLa Cells, 1:500-1:5000; IHC: Human prostate cancer, 1:20-1:200

Type: Primary
Antigen: ZNF266
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human