Anti-MMAB Rabbit Polyclonal Antibody
Catalog # 10089-660
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Methylmalonic aciduria type B
- Antigen symbol:MMAB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:ATR|cob|CFAP23|cblB
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10089-660
- Supplier No.:11137-1-AP
Specifications
About this item
The MMAB gene encodes cob(I)alamin adenosyltransferase, which catalyzes the final step in the synthesis of the coenzyme adenosylcobalamin (AdoCbl). AdoCbl is a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. The deduced 250-amino acid protein has a calculated molecular mass of 27.3 kD.
Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human ovary tumor Tissue, 1:20-1:200
Type: Primary
Antigen: MMAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse