Order Entry
Canada
ContactUsLinkComponent
 
Anti-TRIM11 Rabbit Polyclonal Antibody
Catalog # 10096-312
Supplier:  Proteintech
undefined
Anti-TRIM11 Rabbit Polyclonal Antibody
Catalog # 10096-312
Supplier:  Proteintech
Supplier Number:  10851-1-AP-150UL

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tripartite Motif Containing 11
  • Antigen symbol:
    TRIM11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Size:
    150 µL
  • Antigen synonyms:
    BIA1|RNF92
  • Storage buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Shipping temperature:
    Store at -20°C
  • Immunogen:
    Recombinant Protein
  • Purification:
    Antigen affinity Purification
  • Cat. no.:
    10096-312
  • Supplier No.:
    10851-1-AP

Specifications

About this item

TRIM11(Tripartite motif-containing protein 11) is also named as RNF92 and Belongs to the TRIM/RBCC family. It is a member of the tripartite-motif-containing protein family and is known to destabilize humanin, an inhibitor of Alzheimer-like neuronal insults. This protein has 3 isoforms produced by alternative splicing.

Western Blot: HEK-293 Cells, 1:200-1:1000; IHC: Human ovary tumor Tissue, 1:20-1:200

Type: Primary
Antigen: TRIM11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human