Specifications
- Antibody type:Primary
- Antigen name:BCL2-Like 1
- Antigen symbol:BCL2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:Regulatory Subunit 52|Apoptosis Regulator Bcl-X|BCLXS|Protein Phosphatase 1|BCL2L|PPP1R52
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10083-558
- Supplier No.:10783-1-AP
Specifications
About this item
BCL2L1 is a member of he BCL-2 protein family. BCL2L1 is expressed as two distinct isoforms, Bcl-xL and Bcl-xS, and is located at the outer mitochondrial membrane. The Bcl-XL isoform is a 233 amino acid protein, acting as an apoptotic inhibitor. Bcl-XL can forms heterodimers with BAX, BAK or BCL2, and the heterodimerization with BAX does not seem to be required for anti-apoptotic activity. The Bcl-XS isoform is a shorter variant that is 178 amino acids in length and lacks a 63 amino acid region (amino acids 126-188), acting as an apoptotic activator. This antibody can recognize Bcl-XL, Bcl-XS and Bcl-X(Beta).
IHC: Human Colon cancer Tissue, 1:20-1:200; IP:K-562 Cells, 1:200-1:2000; Western Blot:C6 Cells, 1:500-1:5000; IF:NIH/3T3 Cells, 1:10-1:100
Type: Primary
Antigen: BCL2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat