Anti-VAMP4 Rabbit Polyclonal Antibody
Catalog # 10096-908
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Vesicle-Associated Membrane Protein 4
- Antigen symbol:VAMP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:VAMP24|VAMP-4
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10096-908
- Supplier No.:10738-1-AP
Specifications
About this item
Western Blot:SH-SY5Y Cells, 1:200-1:2000
Type: Primary
Antigen: VAMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat