Anti-SULT1C2 Rabbit Polyclonal Antibody
Catalog # 10095-386
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Sulfotransferase family Cytosolic 1C Member 2
- Antigen symbol:SULT1C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:ST1C2|ST1C1|humSULTC2|SULT1C1
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10095-386
- Supplier No.:10662-1-AP
Specifications
About this item
SULT1C2, also named as SULT1C1, ST1C2, ST1C1, and humSULTC2, belongs to the sulfotransferase 1 family. It is a sulfotransferase enzymes that catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. SULT1C2 may be involved in the activation of carcinogenic hyroxylamines. It is responsible for transferring a sulfo moiety from PAPS to phenol-containing compounds.
Western Blot: Human Stomach Tissue, 1:500-1:5000; IHC: Human prostate cancer, 1:20-1:200
Type: Primary
Antigen: SULT1C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human