Anti-SNCB Rabbit Polyclonal Antibody
Catalog # 10094-926
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Synuclein, Beta
- Antigen symbol:SNCB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:SNCB|beta-synuclein|Beta-synuclein.
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10094-926
- Supplier No.:10498-1-AP
Specifications
About this item
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human breast cancer Tissue, 1:10-1:100; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A
Type: Primary
Antigen: SNCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat