Anti-HSPBP1 Rabbit Polyclonal Antibody
Catalog # 10087-634
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Heat Shock Protein-Binding Protein 1
- Antigen symbol:HSPBP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:Hsp70-Binding Protein 2|FES1|Hsp70-Interacting Protein 2|Hsp70 Interacting Protein|HSPA (Heat Shock 70kDa) Binding Protein|Cytoplasmic Cochaperone 1
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10087-634
- Supplier No.:10211-1-AP
Specifications
About this item
Hsp70-interacting protein (HSPBP1)is a protein of approximately 40 kilodaltons detected in cell extracts. Its mRNA is present in all tissues analyzed but is most abundant in heart and skeletal muscle. HSPBP1 binds the ATPase domain of Hsp70 and inhibits approximately 90% of the Hsp40-activated Hsp70 ATPase activity by preventing ATP binding to Hsp70.
Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Colon cancer, 1:20-1:200
Type: Primary
Antigen: HSPBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse