Specifications
- Antibody type:Primary
- Antigen symbol:DIDO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10085-858
- Supplier No.:10183-1-AP
Specifications
About this item
DIDO1, also termed DATF-1, C20orf158, DATF1, KIAA0333 and DIO-1, is a putative transcription factor, weakly pro-apoptotic when overexpressed. It is a tumor suppressor. In mice, DIDO1 is upregulated by apoptotic signals and encodes a cytoplasmic protein that translocates to the nucleus upon apoptotic signal activation. When overexpressed, the mouse protein induced apoptosis in cell lines growing in vitro. The human DIDO1 is similar to the mouse gene and therefore is thought to be involved in apoptosis. The immunogen of this antibody is the C-terminal of isoform 2 of DATF1(DIDO1).
Western Blot: Human Placenta Tissue, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A
Type: Primary
Antigen: DIDO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat