Anti-SPIN2B Rabbit Polyclonal Antibody
Catalog # 10095-096
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:Spindlin 2A
- Antigen symbol:SPIN2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10095-096
- Supplier No.:10107-2-AP
Specifications
About this item
Western Blot: Mouse Brain Tissue, 1:200-1:2000; IHC: Human breast cancer, 1:20-1:200
Type: Primary
Antigen: SPIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat