Anti-SNAPIN Rabbit Polyclonal Antibody
Catalog # 10094-922
Supplier: Proteintech
Specifications
- Antibody type:Primary
- Antigen name:SNAP-associated Protein
- Antigen symbol:SNAPIN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:150 µL
- Antigen synonyms:BLOC1S7|BLOS7|SNAP25BP|SNAPAP
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Shipping temperature:Store at -20°C
- Immunogen:Recombinant Protein
- Purification:Antigen affinity Purification
- Cat. no.:10094-922
- Supplier No.:10055-1-AP
Specifications
About this item
Snapin, a protein of relative molecular weight of 15kd, is implicated in neurotransmission by binding to SNAP-25. Snapin was enriched in neurons and exclusively located on synaptic vesicle membrane, which may be a PKA target for modulating transmitter release through the cAMP-dependent signal-transduction pathway. This anitbody can recognize the 15-18kd(Monomer) and 30-36kd(Dimer) forms of SNAPIN.
Western Blot:rat Brain Tissue, 1:200-1:1000; IF: HepG2 Cells, 1:20-1:200; IHC: Human Brain Tissue, 1:20-1:200; FC: HepG2 Cells, N/A
Type: Primary
Antigen: SNAPIN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat