Order Entry
Canada
Orders LinkContactUsLinkComponent
Difco™ A-1 Medium, BD Diagnostics

Difco™ A-1 Medium, BD Diagnostics

A-1 Medium, 500 g
Catalog # CA90000-988
Supplier:   BD
Supplier #:  218231
Difco™ A-1 Medium, BD Diagnostics
Catalog # CA90000-988
Supplier:  BD
CAS Number:  
Difco™ A-1 Medium, BD Diagnostics

A-1 Medium, 500 g

CAS Number:  
Price
274,00 $
Log in for availability
Each (500g)
Product Details & Documents

A-1 medium for the determination of fecal coliforms in water and foods.

Specifications

Specifications

  • Description:
    A-1 Medium
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.