Order Entry
Canada
ContactUsLinkComponent
Rabbit Affinity Purification Resin, ImmunoReagents
Rabbit Affinity Purification Resin, ImmunoReagents
Catalog # 10148-982
Supplier:  Immunoreagents
CAS Number:  
undefined
Rabbit Affinity Purification Resin, ImmunoReagents
Catalog # 10148-982
Supplier:  Immunoreagents
Supplier Number:  GE-015
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Purified Rabbit IgG Coupled to 4% Agarose Beads
  • Size:
    2 mL
  • Storage temperature:
    2 to 8°C, Do Not Freeze
  • Cat. no.:
    10148-982
  • Supplier No.:
    IR1014

Specifications

About this item

ImmunoReagents Rabbit IgG Affinity Prep resins are prepared using highly purified species specific immunoglobulins

Immunoglobulins demonstrate a single arc when analyzed against normal serum proteins by IEP. The resins provided comprise of immunoglobulins covalently bound to a 4% agarose bead utilizing strong and stable hydrazide/aldehyde group chemistry. The Affinity Prep resins have 8-10 mgs of antibodies per/ml of resin, with a 24 month shelf life reagents.

ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulin s from various species cross absorbed to provide high specificity.

This resin is suitable for removal of proteins recognizing Rabbit IgG from liquid solutions, including traditional Immunoabsorptions and Immunoprecipitation protocols.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002
Ordering information: ImmunoReagents products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles. Additionally, products can by customized based on specific assay requirements. Contact your Avantor representative for more information.