Order Entry
ContactUsLinkComponent
117 results for Filters

You searched for: Syringe Filters

Syringe Filters

Syringe filters are attached to the end of a syringe to remove particles from a sample prior to analysis. Filtering liquids, the single-use devices force liquids through the filter either when fluids are initially drawn or delivered. Circular shaped in diameters that fit common syringe sizes, the cartridges remove fragments that would otherwise damage narrow bore equipment. Certified for low levels of UV-absorbing extractables, the syringe filters purify liquids for reproducible results.

Sort by
VWR® Syringe Filters

VWR® Syringe Filters

Supplier: VWR International

Typical applications are clarification, sterile filtration, sample preparation and sterile venting.

Expand 40 Items
Loading...
J.T.Baker®, Syringe Filters

J.T.Baker®, Syringe Filters

Supplier: Avantor

These syringe filters are specifically designed for chromatography sample preparation applications and optimized to provide the most consistent results with less extractables.

Expand 92 Items
Loading...
Acrodisc® Syringe Filters, 13 mm, Cytiva (Formerly Pall Lab)

Acrodisc® Syringe Filters, 13 mm, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

These 13 mm syringe filters offer an available prefiltration layer and wide choice of filter media.

Expand 25 Items
Loading...
ValuPrep Syringe Filters

ValuPrep Syringe Filters

Supplier: Cytiva

ValuPrep syringe filters offer the confidence of Pall membranes in economic 1000 packs.

Expand 19 Items
Loading...
Acrodisc® Ion Chromatography Syringe Filters, Cytiva (Formerly Pall Lab)

Acrodisc® Ion Chromatography Syringe Filters, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.

Expand 10 Items
Loading...
Acrodisc® Syringe Filters, 4 mm, Cytiva (Formerly Pall Lab)

Acrodisc® Syringe Filters, 4 mm, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

These syringe filters are ideal for filtration of gas and organic solvents.

Expand 1 Items
Loading...
Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)

Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.

Expand 31 Items
Loading...
Acrodisc® Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)

Acrodisc® Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

These 25 mm syringe filters offer an available prefiltration layer and wide choice of filter media.

Expand 20 Items
Loading...
Acrodisc® WBC (White Blood Cell) Syringe Filter, Cytiva (Formerly Pall Lab)

Acrodisc® WBC (White Blood Cell) Syringe Filter, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

The Acrodisc® WBC premium syringe filter is ideal for use in the capture and recovery of leukocytes and the separation of red blood cells. Acrodisc® WBC provides a simple and efficient method to isolate blood cells used in a variety of research applications including: Antigen specificity and modulation studies, immune receptor analysis, receptor protein studies, genotyping, gene expression, cell therapy.

Expand 2 Items
Loading...
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)

Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

The new Acrodisc® One and Acrodisc® PSF premium syringe filters combine the benefits of a multi-layered prefiltration device with an automation-certified filter housing.

Expand 31 Items
Loading...
Acrodisc® Syringe Filters, DMSO-Safe, Sterile, Cytiva (Formerly Pall Lab)

Acrodisc® Syringe Filters, DMSO-Safe, Sterile, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

These 25mm filters are ideal for the sterilization of media used for cell cryopreservation.

Expand 1 Items
Loading...
Acrodisc® Syringe Filters for Prefiltration, Cytiva (Formerly Pall Lab)

Acrodisc® Syringe Filters for Prefiltration, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

For prefiltering heavily contaminated aqueous or solvent based samples.

Expand 17 Items
Loading...
Acrodisc® Sterile Syringe Filters for Scale Up, Cytiva (Formerly Pall Lab)

Acrodisc® Sterile Syringe Filters for Scale Up, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

These 25 mm filters are ideal for scale up.

Expand 5 Items
Loading...
Acrodisc® Syringe Filters with Versapor® Membranes, Cytiva (Formerly Pall Lab)

Acrodisc® Syringe Filters with Versapor® Membranes, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

For use in cell culture and other sensitive biological research.

Expand 3 Items
Loading...
Acrodisc® Units with Mustang® E Membrane, Cytiva (Formerly Pall Lab)

Acrodisc® Units with Mustang® E Membrane, Cytiva (Formerly Pall Lab)

Supplier: Cytiva

Disposible, single-use units remove endotoxins in water, buffers, neutral sugars, and certain biological solutions.

Expand 1 Items
Loading...
Nalgene® Syringe Filters, PTFE, 13 mm, Thermo Scientific

Nalgene® Syringe Filters, PTFE, 13 mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

Thermo Scientific™ Nalgene™ 13 mm Ø syringe Filters with PTFE membranes with filter solvents.

Expand 2 Items
Loading...
Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific

Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

Filter sample volumes of 10 to 100 ml with these 25 mm Ø syringe filters.

Expand 8 Items
Loading...
Target® Syringe Filters, Glass Microfiber, National Scientific™

Target® Syringe Filters, Glass Microfiber, National Scientific™

Supplier: Thermo Scientific

Glass microfiber syringe filters are ideal for dissolution testing.

Expand 2 Items
Loading...
Nalgene® Syringe Filters, Nylon, 25mm, Thermo Scientific

Nalgene® Syringe Filters, Nylon, 25mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

These 25 mm Ø Nylon syringe filters are ideal for filtering samples and mobile phases used in GC and HPLC.

Expand 4 Items
Loading...

Target® Syringe Filters, (PES) Membrane, National Scientific™

Supplier: Thermo Scientific

These syringe filters are used for ion chromatography, tissue culture filtration and filtration of proteins and nucleic acids, as well as for filtration of high-temperature liquids.

Expand 2 Items
Loading...
Nalgene® Syringe Filters, Polyethersulfone, 13 mm, Thermo Scientific

Nalgene® Syringe Filters, Polyethersulfone, 13 mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

Thermo Scientific™ Nalgene™ 13 mm Ø syringe filters with PES (polyethersulfone) membrane with filter aqueous solutions and media.

Expand 1 Items
Loading...
Syringe Filters, Color Coded, MicroSolv Technology

Syringe Filters, Color Coded, MicroSolv Technology

Supplier: MicroSolv Technology Corp

Color coded low volume syringe filters for HPLC, CE or dissolution testing.

Expand 54 Items
Loading...
Nalgene® Syringe Filters, PTFE, 25 mm, Thermo Scientific

Nalgene® Syringe Filters, PTFE, 25 mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

Filter solvents with Thermo Scientific™ Nalgene™ 25 mm Ø syringe filters with PTFE membranes.

Expand 2 Items
Loading...
Target® Syringe Filters, Regenerated Cellulose, National Scientific™

Target® Syringe Filters, Regenerated Cellulose, National Scientific™

Supplier: Thermo Scientific

Hydrophilic regenerated cellulose membrane with low extractables and lowest nonspecific binding for biological applications or problem pharmaceutical samples.

Expand 2 Items
Loading...
Minisart® SRP Syringe Filters, Sartorius

Minisart® SRP Syringe Filters, Sartorius

Supplier: Sartorius

Minisart® SRP with chemically inert hydrophobic PTFE is suitable for venting as well as harsh solvents with a pH of 1 to 14.

Expand 19 Items
Loading...
Nalgene® Syringe Filters, Cellulose Acetate, 4 mm, Thermo Scientific

Nalgene® Syringe Filters, Cellulose Acetate, 4 mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

For cold sterilization of aqueous solutions, biological or immunological samples, and cell culture media components.

Expand 2 Items
Loading...
Corning® Plastic Syringe Filters, Corning

Corning® Plastic Syringe Filters, Corning

Supplier: Corning

Syringe filters are available with a choice of membrane type: Polyethersulfone (PES) for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA), with or without prefilter, for lowest protein binding; Polytetrafluoroethylene (PTFE) for chemical resistance; regenerated cellulose (RC) – the best choice for DMSO compatibility; and nylon (NY) which is hydrophilic, surfactant-free and lowest extractable.

Expand 11 Items
Loading...
Target2™ Nylon Syringe Filters, Thermo Scientific

Target2™ Nylon Syringe Filters, Thermo Scientific

Supplier: NATIONAL SCIENTIFIC MX

Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.

Expand 10 Items
Loading...
Nalgene® Syringe Prefilter, Glass Fiber, 25mm, Thermo Scientific

Nalgene® Syringe Prefilter, Glass Fiber, 25mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

Clarify highly viscous solutions and remove large particulates from suspensions using this syringe prefilter.

Expand 1 Items
Loading...
Nalgene® Syringe Filters, Polyethersulfone, 25mm, Thermo Scientific

Nalgene® Syringe Filters, Polyethersulfone, 25mm, Thermo Scientific

Supplier: Thermo Fisher Scientific

Filter aqueous solutions and media with Thermo Scientific™ Nalgene™ 25 mm Ø Syringe Filters with PES (polyethersulfone) membrane.

Expand 2 Items
Loading...
Sort by
Recommended for You