Order Entry
Canada
ContactUsLinkComponent
Whatman™ GV 050 Series Glass Vacuum Filtration Device, Whatman products (Cytiva)
Whatman™ GV 050 Series Glass Vacuum Filtration Device, Whatman products (Cytiva)
Supplier:  Cytiva
Certificates
undefined
Whatman™ GV 050 Series Glass Vacuum Filtration Device, Whatman products (Cytiva)
Supplier:  Cytiva

Specifications

  • Catalog No:
  • CA11012-930
  • Capacity:
  • 250 ml
  • Filters:
  • 47/50 mm
    12.5 cm² surface
  • Filter size:
  • 80 mm dia.
  • Filtration area:
  • 12.5 cm²
  • Pre-filter:
  • 40 mm dia.
  • Funnel capacity:
  • 250 ml
  • Height:
  • 225/450 mm (Height without/with Erlenmeyer flask, diameter without clamp and hose coupling)
  • Application:
  • Foodstuffs (e.g., ice cream)
    Beverages (e.g., residues in beer)
    Pharmaceutical and cosmetic industries
    Water and wastewater
    Residue analytics and precipitation analysis
    Contamination tests (e.g., in electroplating)
    Microbiological, biochemical, and hydrobiological detection
    Radiochemical tests
    Particle analysis in sensitive areas of electronics, aviation, and space travel

Specifications

Frequently Bought Together