Plain Reusable Centrifuge Tubes with Screw Caps, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Threaded ungraduated conical centrifuge tube with a PTFE/rubber-lined black phenolic screw cap.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Ordering information: The 15 ml tubes fit IEC Shield Number 302. The 50 ml tubes fit IEC Shield Number 320.