Whatman™ Polyester Drain Discs
About this item
The polyester drain disc is binder free and has a thickness of 100 µm.
- Flat surface to eliminate filter tearing or rupturing
- Used as a separator between membrane layers in serial stack filtration applications
This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.
5 Options Available Below
Product Details & Documents
The polyester drain disc is binder free and has a thickness of 100 µm.
- Flat surface to eliminate filter tearing or rupturing
- Used as a separator between membrane layers in serial stack filtration applications
This chemically inert support disc is available in a variety of diameters for use in a range of devices. With Application in General laboratory microfiltration, Quality control and sterility testing, Removal of particulates from HPLC solvents, Tissue culture media filtration.
Product Family Options
Product Information
- ØThicknessAvailabilityPrice
- 15000-602Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories10 mm100 µmDiscontinued-Supplier Number: 230300
- CA15000-602Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories10 mm100 µmLog in for availabilityPack of 100$110.60Supplier Number: 230300
- CA11013-694Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories22 mm100 µmLog in for availabilityPack of 100$202.65Supplier Number: 230500
- 10035-598Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories22 mm100 µmDiscontinued-Supplier Number: 230500
- 28156-243Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories25 mm100 µmDiscontinued-Supplier Number: 230600
- CA28156-243Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories25 mm100 µmLog in for availabilityPack of 100$122.37Supplier Number: 230600
- CA11012-804Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories37 mm100 µmLog in for availabilityPack of 100$222.53Supplier Number: 230800
- 10035-600Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories37 mm100 µmDiscontinued-Supplier Number: 230800
- CA11013-696Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories47 mm100 µmLog in for availabilityPack of 100$258.81Supplier Number: 231100
- 10035-602Polyester drain disc, PE, membrane filter accessoriesPolyester drain disc, PE, membrane filter accessories47 mm100 µmDiscontinued-Supplier Number: 231100