Sodium hydroxide
100 g
Supplier: G-Biosciences
Supplier #: S047
Jump to:
Product Details & Documents
Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Boiling Pt:1390 °C (1013 hPa)
- Melting Pt:323 °C
- Density:2.13 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1823
- ADR:8,II
- REACH:01-2119457892-27
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specifications
- Size:100 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



