Blot Washing Buffer, Azure Biosystems
Catalog # 10147-338
Supplier: Azure Biosystems
Product available on GSA Advantage®
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:10X Blot Washing Buffer
- Environmentally Preferable:
- Cat. no.:10147-338
- Volume:500 ml
- Supplier No.:AC2113
Specifications
About this item
Azure Blot Washing Buffer can be used with any Western blotting system.
- Can be used with any Western blotting system
- Provided as a 10X concentrate
- Can be used with all chemiluminescent HRP substrates
Azure Blot Washing Buffer can be used with any Western blotting system, including all chemiluminescent HRP substrates and visible fluorescent Western blotting kits. Provided as a 10X concentrate.