Integra® EC, Extended Cuff Blue Nitrile Gloves, Microflex®
Supplier: Microflex
Certificates
About this item
Integra EC is a durable, 8-millimeter glove for tough jobs where extra durability or safety is needed.
- Ambidextrous
- Non sterile
- Beaded cuffs
- Durable and extra thick
The added thickness of these gloves gives strength and durability. Great protection against bloodborne pathogens. Extra thickness and extended cuff provide extra protection. Gloves are blue, nonsterile, and ambidextrous. They feature a beaded cuff and are fully textured.
Specifications
- Catalog No:
- CA10030-242
- CA10030-246
- CA10030-248
- CA10030-250
- CA10030-252
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Cuff thickness:
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- Finger thickness:
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- Palm thickness:
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil