Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant IFN
Mouse Recombinant IFN
Supplier:  IBI Scientific
Certificates
Mouse Recombinant IFN
Supplier:  IBI Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Gene ID:
    15978
  • Endotoxin-free:
    Yes
  • Protein/peptide name:
    IFN

Specifications

Frequently Bought Together