Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant TNF alpha
Mouse Recombinant TNF alpha
Supplier:  IBI Scientific
Certificates
Mouse Recombinant TNF alpha
Supplier:  IBI Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Mouse
  • Gene ID:
    21926
  • Endotoxin-free:
    Yes
  • Protein/peptide name:
    TNF alpha

Specifications

Frequently Bought Together